missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoroaniline 98.0+%, TCI America™

Catalog No. F010125G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F010125G 25 g
F0101250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F010125G Supplier TCI America Supplier No. F010125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 372-19-0
Molecular Formula C6H6FN
Molecular Weight (g/mol) 111.119
MDL Number MFCD00007758
InChI Key QZVQQUVWFIZUBQ-UHFFFAOYSA-N
Synonym m-fluoroaniline, 1-amino-3-fluorobenzene, 3-fluoranilin, benzenamine, 3-fluoro, 3-fluorobenzenamine, 3-fluoro-phenylamine, aniline, 3-fluoro, 3-fluorophenylamine, aniline, m-fluoro, 3-fluoranilin czech
PubChem CID 9742
ChEBI CHEBI:27873
IUPAC Name 3-fluoroaniline
SMILES C1=CC(=CC(=C1)F)N

Specifications

Color Yellow
Boiling Point 186°C
Quantity 25 g
UN Number 2941
Formula Weight 111.12
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Fluoroaniline
EINECSNumber (3)-0222
RTECSNumber BY1400000
TSCA No