missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Fluoroaniline, 98+%
Description
3-Fluoroaniline is used in the preparation of acetic acid-(3-fluoro-anilide by reacting with acetic acid anhydride. Further, it serves as a reagent in the preparation of radamide analogs as Grp94 inhibitors, which has potential to be antitumor agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Fluoroaniline is used in the preparation of acetic acid-(3-fluoro-anilide by reacting with acetic acid anhydride. Further, it serves as a reagent in the preparation of radamide analogs as Grp94 inhibitors, which has potential to be antitumor agents.
Solubility
Miscible with ethanol and ethyl ether. Slightly miscible with chloroform and water.
Notes
Incompatible with acids, acid chlorides, acid anhydrides, chloroformates and strong oxidizing agents.
3-Fluoroaniline is used in the preparation of acetic acid-(3-fluoro-anilide by reacting with acetic acid anhydride. Further, it serves as a reagent in the preparation of radamide analogs as Grp94 inhibitors, which has potential to be antitumor agents.
Solubility
Miscible with ethanol and ethyl ether. Slightly miscible with chloroform and water.
Notes
Incompatible with acids, acid chlorides, acid anhydrides, chloroformates and strong oxidizing agents.
Specifications
Specifications
| Density | 1.156 |
| Boiling Point | 186°C to 187°C |
| Flash Point | 77°C (170°F) |
| Odor | Recognizable |
| Refractive Index | 1.544 |
| Quantity | 25 g |
| UN Number | UN2941 |
| Beilstein | 1305471 |
| Solubility Information | Miscible with ethanol and ethyl ether. Slightly miscible with chloroform and water. |
| Formula Weight | 111.12 |
| Show More |
Safety and Handling
| Hazard Category | H227-H302+H312+H332-H315-H318-H335 |
| Hazard Statement | GHS H Statement H331-H373-H227-H302-H312-H315-H319 Toxic if inhaled. May cause damage to organs through prolonged or repeated exposure. Combustible liquid. Harmful if swallowed. Harmful in contact with skin. Causes skin irritation. Causes serious eye irritation. |
| Precautionary Statement | P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: FLUOROANILINES |
| EINECSNumber | 206-747-0 |
| RTECSNumber | BY1400000 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only