missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-p-cresol 98.0+%, TCI America™

Catalog No. F04815G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F04815G 5 g
F04811G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F04815G Supplier TCI America Supplier No. F04815G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 452-78-8
Molecular Formula C7H7FO
Molecular Weight (g/mol) 126.13
MDL Number MFCD00143164
InChI Key GJOOCAXPERKNMN-UHFFFAOYSA-N
Synonym 3-fluoro-p-cresol, 3-fluoro-4-methyl-phenol, 2-fluoro-4-hydroxytoluene, phenol, 3-fluoro-4-methyl, pubchem1499, 3-fluoro-4-methyl phenol, acmc-209k1q, 3-fluoranyl-4-methyl-phenol, ksc494a2d
PubChem CID 2774613
IUPAC Name 3-fluoro-4-methylphenol
SMILES CC1=C(C=C(C=C1)O)F

Specifications

Boiling Point 196°C
Quantity 5 g
Formula Weight 126.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 3-Fluoro-p-cresol