accessibility menu, dialog, popup

UserName

CH3OHF

CAS RN 452-78-8

IUPAC Name: 3-fluoro-4-methylphenol
Synonyms: 3-fluoro-4-methylphenol
Molecular Weight (g/mol): 126.13
Molecular Formula: C7H7FO
InChi Key: GJOOCAXPERKNMN-UHFFFAOYSA-N
SMILES: CC1=C(F)C=C(O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results