missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-p-anisaldehyde 98.0+%, TCI America™

Catalog No. p-7129710
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F04801G 1 g
F04805G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F04801G Supplier TCI America Supplier No. F04801G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 351-54-2
Molecular Formula C8H7FO2
Molecular Weight (g/mol) 154.14
InChI Key SOQCZBSZZLWDGU-UHFFFAOYSA-N
Synonym 3-fluoro-p-anisaldehyde, 3-fluoro-4-methoxy-benzaldehyde, 3-fluoro-4-anisaldehyde, benzaldehyde, 3-fluoro-4-methoxy, 3-fluoro-4-methoxy benzaldehyde, 3-fluoroanisaldehyde, 3-fluoro-anisaldehyde, pubchem1448, acmc-209idn, 2-fluoro-4-formylanisole
PubChem CID 67696
IUPAC Name 3-fluoro-4-methoxybenzaldehyde
SMILES COC1=C(C=C(C=C1)C=O)F

Specifications

Color White-Yellow
Boiling Point 132°C
Quantity 1 g
Formula Weight 154.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 3-Fluoro-p-anisaldehyde