accessibility menu, dialog, popup

UserName

CAS RN 351-54-2

IUPAC Name: 3-fluoro-4-methoxybenzaldehyde
Synonyms: 3-fluoro-4-methoxybenzaldehyde
Molecular Weight (g/mol): 154.14
Molecular Formula: C8H7FO2
InChi Key: SOQCZBSZZLWDGU-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=O)C=C1F
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)