missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Fluoro-L-tyrosine, 97%, Thermo Scientific™
Chemical Identifiers
| CAS | 7423-96-3 |
|---|---|
| Molecular Formula | C9H10FNO3 |
| Molecular Weight (g/mol) | 199.181 |
| MDL Number | MFCD00002607 |
| InChI Key | VIIAUOZUUGXERI-ZETCQYMHSA-N |
| Synonym | 3-fluoro-l-tyrosine, s-2-amino-3-3-fluoro-4-hydroxyphenyl propanoic acid, 3-fluorotyrosine, 3-fluorotyrosine, l, h-tyr 3-f-oh, unii-174nrg3m2x, s-3-fluorotyrosine, 3'-fluoro-l-tyrosine, 2s-2-amino-3-3-fluoro-4-hydroxyphenyl propanoic acid, 3-fluoro-tyrosine |
| PubChem CID | 643330 |
| ChEBI | CHEBI:46534 |
| IUPAC Name | (2S)-2-amino-3-(3-fluoro-4-hydroxyphenyl)propanoic acid |
| SMILES | C1=CC(=C(C=C1CC(C(=O)O)N)F)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AAL16775MF
|
N/A | 50mg |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications3-Fluoro-L-tyrosine is used as pharma and intermediates.
Solubility
Soluble in water (9.8 g/L).
Notes
Keep container tightly closed. Store away from oxidizing agents.
Chemical Identifiers
| 7423-96-3 | |
| 199.181 | |
| VIIAUOZUUGXERI-ZETCQYMHSA-N | |
| 643330 | |
| (2S)-2-amino-3-(3-fluoro-4-hydroxyphenyl)propanoic acid |
| C9H10FNO3 | |
| MFCD00002607 | |
| 3-fluoro-l-tyrosine, s-2-amino-3-3-fluoro-4-hydroxyphenyl propanoic acid, 3-fluorotyrosine, 3-fluorotyrosine, l, h-tyr 3-f-oh, unii-174nrg3m2x, s-3-fluorotyrosine, 3'-fluoro-l-tyrosine, 2s-2-amino-3-3-fluoro-4-hydroxyphenyl propanoic acid, 3-fluoro-tyrosine | |
| CHEBI:46534 | |
| C1=CC(=C(C=C1CC(C(=O)O)N)F)O |
Specifications
| 7423-96-3 | |
| MFCD00002607 | |
| 3204802 | |
| Soluble in water (9.8g/L). | |
| C1=CC(=C(C=C1CC(C(=O)O)N)F)O | |
| 199.181 | |
| CHEBI:46534 | |
| −7° (c=1 in 1M HCl) | |
| 3-Fluoro-L-tyrosine |
| C9H10FNO3 | |
| UN2811 | |
| 3-fluoro-l-tyrosine, s-2-amino-3-3-fluoro-4-hydroxyphenyl propanoic acid, 3-fluorotyrosine, 3-fluorotyrosine, l, h-tyr 3-f-oh, unii-174nrg3m2x, s-3-fluorotyrosine, 3'-fluoro-l-tyrosine, 2s-2-amino-3-3-fluoro-4-hydroxyphenyl propanoic acid, 3-fluoro-tyrosine | |
| VIIAUOZUUGXERI-ZETCQYMHSA-N | |
| (2S)-2-amino-3-(3-fluoro-4-hydroxyphenyl)propanoic acid | |
| 643330 | |
| 199.18 | |
| 97% |
Safety and Handling
GHS H Statement
H300
Fatal if swallowed.
P264b-P270-P301+P310-P330-P501c
H301
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only