missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-L-tyrosine, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals L16775MF

Quantity 50mg

 View more versions of this product

Catalog No. AAL16775MF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3-Fluoro-L-tyrosine is used as pharma and intermediates.

Solubility
Soluble in water (9.8 g/L).

Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications

Chemical Identifiers

7423-96-3
199.181
VIIAUOZUUGXERI-ZETCQYMHSA-N
643330
(2S)-2-amino-3-(3-fluoro-4-hydroxyphenyl)propanoic acid
C9H10FNO3
MFCD00002607
3-fluoro-l-tyrosine, s-2-amino-3-3-fluoro-4-hydroxyphenyl propanoic acid, 3-fluorotyrosine, 3-fluorotyrosine, l, h-tyr 3-f-oh, unii-174nrg3m2x, s-3-fluorotyrosine, 3'-fluoro-l-tyrosine, 2s-2-amino-3-3-fluoro-4-hydroxyphenyl propanoic acid, 3-fluoro-tyrosine
CHEBI:46534
C1=CC(=C(C=C1CC(C(=O)O)N)F)O

Specifications

3-Fluoro-L-tyrosine
C9H10FNO3
UN2811
3-fluoro-l-tyrosine, s-2-amino-3-3-fluoro-4-hydroxyphenyl propanoic acid, 3-fluorotyrosine, 3-fluorotyrosine, l, h-tyr 3-f-oh, unii-174nrg3m2x, s-3-fluorotyrosine, 3'-fluoro-l-tyrosine, 2s-2-amino-3-3-fluoro-4-hydroxyphenyl propanoic acid, 3-fluoro-tyrosine
VIIAUOZUUGXERI-ZETCQYMHSA-N
(2S)-2-amino-3-(3-fluoro-4-hydroxyphenyl)propanoic acid
50mg
CHEBI:46534
−7° (c=1 in 1M HCl)
7423-96-3
MFCD00002607
3204802
Soluble in water (9.8g/L).
C1=CC(=C(C=C1CC(C(=O)O)N)F)O
199.181
643330
199.18
97%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300
Fatal if swallowed.

P264b-P270-P301+P310-P330-P501c

H301

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

Promo
Expires: 06-30-2027
Get Up to $1,800 in Chemicals with Purchase
Spend $250 or more on Thermo Fisher Scientific chemicals and get up to $1,800 to spend on additional chemicals.

Adding...