accessibility menu, dialog, popup

UserName

(S)ONH2OHHOF

CAS RN 7423-96-3

IUPAC Name: (2S)-2-amino-3-(3-fluoro-4-hydroxyphenyl)propanoic acid
Synonyms: 3-fluorotyrosine
Molecular Weight (g/mol): 199.18
Molecular Formula: C9H10FNO3
InChi Key: VIIAUOZUUGXERI-JLDDOWRYNA-N
SMILES: N[C@@H](CC1=CC=C(O)C(F)=C1)C(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)