Learn More
3-Fluoro-5-(pentafluorothio)aniline, 97%
CAS: 1240256-99-8 | C6H5F6NS | 237.163 g/mol
Supplier: Thermo Scientific Chemicals H3368003
Description
It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.
Solubility
Sparingly soluble in water.(0.26 g/L) (25°C),
Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.
Specifications
| 3-Fluoro-5-(pentafluorothio)aniline | |
| C6H5F6NS | |
| 1 g | |
| 3-fluoro-5-pentafluorosulfur aniline, 3-fluoro-5-pentafluoro-??-sulfanyl aniline, 3-fluoro-5-pentafluorothio aniline | |
| RFXWDLYDFAZNLO-UHFFFAOYSA-N | |
| 3-fluoro-5-(pentafluoro-$l^{6}-sulfanyl)aniline | |
| 66523473 | |
| 97% |
| 1240256-99-8 | |
| MFCD16652430 | |
| UN2811 | |
| Sparingly soluble in water.(0.26g/L) (25°C), | |
| C1=C(C=C(C=C1F)S(F)(F)(F)(F)F)N | |
| 237.163 | |
| 237.16 |
Chemical Identifiers
| 1240256-99-8 | |
| 237.163 | |
| RFXWDLYDFAZNLO-UHFFFAOYSA-N | |
| 66523473 | |
| C1=C(C=C(C=C1F)S(F)(F)(F)(F)F)N |
| C6H5F6NS | |
| MFCD16652430 | |
| 3-fluoro-5-pentafluorosulfur aniline, 3-fluoro-5-pentafluoro-??-sulfanyl aniline, 3-fluoro-5-pentafluorothio aniline | |
| 3-fluoro-5-(pentafluoro-$l^{6}-sulfanyl)aniline |
Safety and Handling
GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only