accessibility menu, dialog, popup

UserName

CAS RN 1240256-99-8

IUPAC Name: 3-fluoro-5-(pentafluoro-λ⁶-sulfanyl)aniline
Synonyms: 3-fluoro-5-(pentafluoro-λ⁶-sulfanyl)aniline
Molecular Weight (g/mol): 237.16
Molecular Formula: C6H5F6NS
InChi Key: RFXWDLYDFAZNLO-UHFFFAOYSA-N
SMILES: NC1=CC(F)=CC(=C1)S(F)(F)(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results