missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-5-(methoxycarbonyl)benzeneboronic acid, 95%, Thermo Scientific Chemicals

Catalog No. p-7035367
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5308403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5308403 Supplier Thermo Scientific Chemicals Supplier No. H5308403

3-Fluoro-5-(methoxycarbonyl)benzeneboronic acid is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Fluoro-5-(methoxycarbonyl)benzeneboronic acid is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 871329-62-3
Molecular Formula C8H8BFO4
Molecular Weight (g/mol) 197.956
MDL Number MFCD07363749
InChI Key VKHJVASTGLPBBL-UHFFFAOYSA-N
Synonym 3-fluoro-5-methoxycarbonyl phenyl boronic acid, 3-fluoro-5-methoxycarbonylphenyl boronic acid, 3-fluoro-5-methoxycarbonyl phenylboronic acid, 3-fluoro-5-methoxycarbonyl benzeneboronic acid, benzoic acid,3-borono-5-fluoro-, 1-methyl ester 9ci, methyl 3-borono-5-fluorobenzoate, acmc-209qg6, ksc657s1f, 3-fluoro-5-methoxycarbonylphenylboronicacid
PubChem CID 44717206
IUPAC Name (3-fluoro-5-methoxycarbonylphenyl)boronic acid
SMILES B(C1=CC(=CC(=C1)F)C(=O)OC)(O)O

Specifications

Melting Point 138°C to 142°C
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 197.96
Percent Purity 95%
Chemical Name or Material 3-Fluoro-5-(methoxycarbonyl)benzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only