CAS RN 871329-62-3
IUPAC Name:
[3-fluoro-5-(methoxycarbonyl)phenyl]boronic acid
Synonyms:
3-fluoro-5-(methoxycarbonyl)phenylboronic acid
Molecular Weight (g/mol):
197.96
Molecular Formula:
C8H8BFO4
InChi Key:
VKHJVASTGLPBBL-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CC(F)=CC(=C1)B(O)O
1
–
1
of
1
results
1
Promotions Available
3-Fluoro-5-(methoxycarbonyl)benzeneboronic acid, 95%, Thermo Scientific Chemicals
3-Fluoro-5-(methoxycarbonyl)benzeneboronic acid, 95%, Thermo Scientific Chemicals
CAS: 871329-62-3 Molecular Formula: C8H8BFO4 Molecular Weight (g/mol): 197.956 MDL Number: MFCD07363749 InChI Key: VKHJVASTGLPBBL-UHFFFAOYSA-N Synonym: 3-fluoro-5-methoxycarbonyl phenyl boronic acid,3-fluoro-5-methoxycarbonylphenyl boronic acid,3-fluoro-5-methoxycarbonyl phenylboronic acid,3-fluoro-5-methoxycarbonyl benzeneboronic acid,benzoic acid,3-borono-5-fluoro-, 1-methyl ester 9ci,methyl 3-borono-5-fluorobenzoate,acmc-209qg6,ksc657s1f,3-fluoro-5-methoxycarbonylphenylboronicacid PubChem CID: 44717206 IUPAC Name: (3-fluoro-5-methoxycarbonylphenyl)boronic acid SMILES: B(C1=CC(=CC(=C1)F)C(=O)OC)(O)O