missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-(trifluoromethoxy)phenol 98.0+%, TCI America™

Catalog No. F10091G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F10091G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F10091G Supplier TCI America Supplier No. F10091G

Chemical Identifiers

CAS 177596-38-2
Molecular Formula C7H4F4O2
Molecular Weight (g/mol) 196.101
MDL Number MFCD06660342
InChI Key UTFSPWRTXCDUIJ-UHFFFAOYSA-N
Synonym 3-fluoro-4-trifluoromethoxy phenol, phenol, 3-fluoro-4-trifluoromethoxy, 1-fluoro-5-hydroxy-2-trifluoromethoxy benzene, acmc-1bxa3, 3-fluoro-4-trifluromethoxyphenol, 3-fluoranyl-4-trifluoromethyloxy phenol
PubChem CID 2783357
IUPAC Name 3-fluoro-4-(trifluoromethoxy)phenol
SMILES C1=CC(=C(C=C1O)F)OC(F)(F)F

Specifications

Quantity 1 g
Formula Weight 196.10
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Fluoro-4-(trifluoromethoxy)phenol