CAS RN 177596-38-2
IUPAC Name:
3-fluoro-4-(trifluoromethoxy)phenol
Synonyms:
3-fluoro-4-(trifluoromethoxy)phenol
Molecular Weight (g/mol):
196.1
Molecular Formula:
C7H4F4O2
InChi Key:
UTFSPWRTXCDUIJ-UHFFFAOYSA-N
SMILES:
OC1=CC(F)=C(OC(F)(F)F)C=C1
1
–
1
of
1
results
1
3-Fluoro-4-(trifluoromethoxy)phenol 98.0+%, TCI America™
CAS: 177596-38-2 Molecular Formula: C7H4F4O2 Molecular Weight (g/mol): 196.101 MDL Number: MFCD06660342 InChI Key: UTFSPWRTXCDUIJ-UHFFFAOYSA-N Synonym: 3-fluoro-4-trifluoromethoxy phenol,phenol, 3-fluoro-4-trifluoromethoxy,1-fluoro-5-hydroxy-2-trifluoromethoxy benzene,acmc-1bxa3,3-fluoro-4-trifluromethoxyphenol,3-fluoranyl-4-trifluoromethyloxy phenol PubChem CID: 2783357 IUPAC Name: 3-fluoro-4-(trifluoromethoxy)phenol SMILES: C1=CC(=C(C=C1O)F)OC(F)(F)F