missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-nitrobenzyl bromide, 97%

Catalog No. AAH2619303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2619303 1 g
AAH2619306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2619303 Supplier Thermo Scientific Chemicals Supplier No. H2619303
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 131858-37-2 | C7H5BrFNO2 | 234.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 131858-37-2
Molecular Formula C7H5BrFNO2
Molecular Weight (g/mol) 234.02
MDL Number MFCD03094237
InChI Key ZOZJSWIXPIVMRU-UHFFFAOYSA-N
Synonym 3-fluoro-4-nitrobenzyl bromide, 4-bromomethyl-2-fluoro-1-nitrobenzene, 4-bromomethyl-2-fluoronitrobenzene, benzene, 4-bromomethyl-2-fluoro-1-nitro, intermediates-zcf02012, acmc-1c6h7, 4-bromomethyl-2-fluoro-1-nitro-benzene, benzene,4-bromomethyl-2-fluoro-1-nitro, 4-bromomethyl-2-fluoranyl-1-nitro-benzene
PubChem CID 2774661
IUPAC Name 4-(bromomethyl)-2-fluoro-1-nitrobenzene
SMILES [O-][N+](=O)C1=C(F)C=C(CBr)C=C1

Specifications

Melting Point 46°C to 48°C
Quantity 1 g
UN Number UN3261
Formula Weight 234.02
Percent Purity 97%
Chemical Name or Material 3-Fluoro-4-nitrobenzyl bromide
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318-H335
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only