accessibility menu, dialog, popup

UserName

BrFNOO

CAS RN 131858-37-2

IUPAC Name: 4-(bromomethyl)-2-fluoro-1-nitrobenzene
Synonyms: 4-(bromomethyl)-2-fluoro-1-nitrobenzene
Molecular Weight (g/mol): 234.02
Molecular Formula: C7H5BrFNO2
InChi Key: ZOZJSWIXPIVMRU-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C(F)C=C(CBr)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results