missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Fluoro-4-nitrobenzaldehyde 97.0+%, TCI America™
$144.39 - $352.31
Chemical Identifiers
| CAS | 160538-51-2 |
|---|---|
| Molecular Formula | C7H4FNO3 |
| Molecular Weight (g/mol) | 169.111 |
| MDL Number | MFCD00968940 |
| InChI Key | BWUIGISQVCIQBT-UHFFFAOYSA-N |
| PubChem CID | 3808120 |
| IUPAC Name | 3-fluoro-4-nitrobenzaldehyde |
| SMILES | C1=CC(=C(C=C1C=O)F)[N+](=O)[O-] |
Chemical Identifiers
| 160538-51-2 | |
| 169.111 | |
| BWUIGISQVCIQBT-UHFFFAOYSA-N | |
| 3-fluoro-4-nitrobenzaldehyde |
| C7H4FNO3 | |
| MFCD00968940 | |
| 3808120 | |
| C1=CC(=C(C=C1C=O)F)[N+](=O)[O-] |
Specifications
| 160538-51-2 | |
| C7H4FNO3 | |
| 1 g | |
| C1=CC(=C(C=C1C=O)F)[N+](=O)[O-] | |
| 169.111 | |
| 169.11 | |
| Crystalline Powder |
| 55°C | |
| MFCD00968940 | |
| BWUIGISQVCIQBT-UHFFFAOYSA-N | |
| 3-fluoro-4-nitrobenzaldehyde | |
| 3808120 | |
| ≥97.0% (GC) | |
| 3-Fluoro-4-nitrobenzaldehyde |
Safety and Handling
TSCA : No