accessibility menu, dialog, popup

UserName

CAS RN 160538-51-2

OFNOO

CAS RN 160538-51-2

IUPAC Name: 3-fluoro-4-nitrobenzaldehyde
Synonyms: 3-fluoro-4-nitrobenzaldehyde
Molecular Weight (g/mol): 169.11
Molecular Formula: C7H4FNO3
InChi Key: BWUIGISQVCIQBT-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C(F)C=C(C=O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results