missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Fluoro-4-methylphenylboronic Acid (contains varying amounts of Anhydride) 97.0+%, TCI America™
$59.65 - $169.63
Chemical Identifiers
| CAS | 168267-99-0 |
|---|---|
| Molecular Formula | C7H8BFO2 |
| Molecular Weight (g/mol) | 153.947 |
| MDL Number | MFCD02683115 |
| InChI Key | WPVBHUUZDFUIJA-UHFFFAOYSA-N |
| Synonym | 3-fluoro-4-methylbenzeneboronic acid, 3-fluoro-4-methylphenyl boronic acid, 3-fluoro-p-tolylboronic acid, 3-fluoro-4-methylphenyl-boronic acid, boronic acid, 3-fluoro-4-methylphenyl, pubchem5124, 3-fluoro-4-methyl-phenyl boronic acid, acmc-209dxx, ksc173s0n |
| PubChem CID | 2782674 |
| IUPAC Name | (3-fluoro-4-methylphenyl)boronic acid |
| SMILES | B(C1=CC(=C(C=C1)C)F)(O)O |
Chemical Identifiers
| 168267-99-0 | |
| 153.947 | |
| WPVBHUUZDFUIJA-UHFFFAOYSA-N | |
| 2782674 | |
| B(C1=CC(=C(C=C1)C)F)(O)O |
| C7H8BFO2 | |
| MFCD02683115 | |
| 3-fluoro-4-methylbenzeneboronic acid, 3-fluoro-4-methylphenyl boronic acid, 3-fluoro-p-tolylboronic acid, 3-fluoro-4-methylphenyl-boronic acid, boronic acid, 3-fluoro-4-methylphenyl, pubchem5124, 3-fluoro-4-methyl-phenyl boronic acid, acmc-209dxx, ksc173s0n | |
| (3-fluoro-4-methylphenyl)boronic acid |
Specifications
| 168267-99-0 | |
| White-Yellow | |
| MFCD02683115 | |
| 3-fluoro-4-methylbenzeneboronic acid, 3-fluoro-4-methylphenyl boronic acid, 3-fluoro-p-tolylboronic acid, 3-fluoro-4-methylphenyl-boronic acid, boronic acid, 3-fluoro-4-methylphenyl, pubchem5124, 3-fluoro-4-methyl-phenyl boronic acid, acmc-209dxx, ksc173s0n | |
| B(C1=CC(=C(C=C1)C)F)(O)O | |
| 153.947 | |
| 153.95 | |
| Crystalline Powder |
| 237°C | |
| C7H8BFO2 | |
| 1 g | |
| WPVBHUUZDFUIJA-UHFFFAOYSA-N | |
| (3-fluoro-4-methylphenyl)boronic acid | |
| 2782674 | |
| ≥97.0% (T) | |
| 3-Fluoro-4-methylphenylboronic Acid (contains varying amounts of Anhydride) |
Safety and Handling
TSCA : No