accessibility menu, dialog, popup

UserName

CH3FBOHHO

CAS RN 168267-99-0

IUPAC Name: (3-fluoro-4-methylphenyl)boronic acid
Synonyms: 3-fluoro-4-methylphenylboronic acid
Molecular Weight (g/mol): 153.95
Molecular Formula: C7H8BFO2
InChi Key: WPVBHUUZDFUIJA-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1F)B(O)O
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)