missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-methylbenzoic Acid 98.0+%, TCI America™

Catalog No. p-7129749
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F05525G 5 g
F055225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F05525G Supplier TCI America Supplier No. F05525G

Chemical Identifiers

CAS 350-28-7
Molecular Formula C8H7FO2
Molecular Weight (g/mol) 154.14
MDL Number MFCD00002490
InChI Key XUQCONCMPCVUDM-UHFFFAOYSA-N
Synonym 3-fluoro-p-toluic acid, 3-fluoro-4-methylbenzoicacid, 3-fluoro-4-methyl-benzoic acid, benzoic acid, 3-fluoro-4-methyl, pubchem1330, acmc-209ich, 4-carboxy-2-fluorotoluene, 3-fluoro4-methylbenzoic acid, ksc223k8t, rarechem al bo 0761
PubChem CID 67687
IUPAC Name 3-fluoro-4-methylbenzoic acid
SMILES CC1=CC=C(C=C1F)C(O)=O

Specifications

Melting Point 173°C
Color White-Yellow
Quantity 5 g
Formula Weight 154.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Fluoro-4-methylbenzoic Acid