CAS RN 350-28-7
CAS RN 350-28-7
IUPAC Name:
3-fluoro-4-methylbenzoic acid
Synonyms:
3-fluoro-4-methylbenzoic acid
Molecular Weight (g/mol):
154.14
Molecular Formula:
C8H7FO2
InChi Key:
XUQCONCMPCVUDM-UHFFFAOYSA-N
SMILES:
CC1=CC=C(C=C1F)C(O)=O
1
–
1
of
1
results
1
3-Fluoro-4-methylbenzoic Acid 98.0+%, TCI America™
CAS: 350-28-7 Molecular Formula: C8H7FO2 Molecular Weight (g/mol): 154.14 MDL Number: MFCD00002490 InChI Key: XUQCONCMPCVUDM-UHFFFAOYSA-N Synonym: 3-fluoro-p-toluic acid,3-fluoro-4-methylbenzoicacid,3-fluoro-4-methyl-benzoic acid,benzoic acid, 3-fluoro-4-methyl,pubchem1330,acmc-209ich,4-carboxy-2-fluorotoluene,3-fluoro4-methylbenzoic acid,ksc223k8t,rarechem al bo 0761 PubChem CID: 67687 IUPAC Name: 3-fluoro-4-methylbenzoic acid SMILES: CC1=CC=C(C=C1F)C(O)=O