missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-methylbenzeneboronic acid, 98%

CAS: 168267-99-0 | C7H8BFO2 | 153.947 g/mol

Supplier:  Thermo Scientific Chemicals B2451206

 View more versions of this product

Catalog No. AAB2451206

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Fluoro-4-methylbenzeneboronic acid
234°C
MFCD02683115
8544594
WPVBHUUZDFUIJA-UHFFFAOYSA-N
(3-fluoro-4-methylphenyl)boronic acid
2782674
98%
168267-99-0
C7H8BFO2
5 g
3-fluoro-4-methylbenzeneboronic acid, 3-fluoro-4-methylphenyl boronic acid, 3-fluoro-p-tolylboronic acid, 3-fluoro-4-methylphenyl-boronic acid, boronic acid, 3-fluoro-4-methylphenyl, pubchem5124, 3-fluoro-4-methyl-phenyl boronic acid, acmc-209dxx, ksc173s0n
B(C1=CC(=C(C=C1)C)F)(O)O
153.947
153.95

Chemical Identifiers

168267-99-0
153.947
WPVBHUUZDFUIJA-UHFFFAOYSA-N
2782674
B(C1=CC(=C(C=C1)C)F)(O)O
C7H8BFO2
MFCD02683115
3-fluoro-4-methylbenzeneboronic acid, 3-fluoro-4-methylphenyl boronic acid, 3-fluoro-p-tolylboronic acid, 3-fluoro-4-methylphenyl-boronic acid, boronic acid, 3-fluoro-4-methylphenyl, pubchem5124, 3-fluoro-4-methyl-phenyl boronic acid, acmc-209dxx, ksc173s0n
(3-fluoro-4-methylphenyl)boronic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only