missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-methylbenzaldehyde 95.0+%, TCI America™

Catalog No. F07925G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F07925G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F07925G Supplier TCI America Supplier No. F07925G
Only null left

Chemical Identifiers

CAS 177756-62-6
Molecular Formula C8H7FO
Molecular Weight (g/mol) 138.141
MDL Number MFCD01631532
InChI Key UFPBMVRONDLOGK-UHFFFAOYSA-N
Synonym 3-fluoro-p-tolualdehyde, 3-fluoro-4-methyl-benzaldehyde, benzaldehyde, 3-fluoro-4-methyl-9ci, pubchem1454, acmc-1c3kx, ksc494i5j, 3-fluoranyl-4-methyl-benzaldehyde, 3-fluoro-4-methylbenzaldehyde, benzaldehyde, 3-fluoro-4-methyl
PubChem CID 2774574
IUPAC Name 3-fluoro-4-methylbenzaldehyde
SMILES CC1=C(C=C(C=C1)C=O)F

Specifications

Color Yellow
Boiling Point 206°C
Quantity 5 g
Formula Weight 138.14
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Fluoro-4-methylbenzaldehyde