CAS RN 177756-62-6
IUPAC Name:
3-fluoro-4-methylbenzaldehyde
Synonyms:
3-fluoro-4-methylbenzaldehyde
Molecular Weight (g/mol):
138.14
Molecular Formula:
C8H7FO
InChi Key:
UFPBMVRONDLOGK-UHFFFAOYSA-N
SMILES:
CC1=CC=C(C=O)C=C1F
1
–
1
of
1
results
1
3-Fluoro-4-methylbenzaldehyde 95.0+%, TCI America™
CAS: 177756-62-6 Molecular Formula: C8H7FO Molecular Weight (g/mol): 138.141 MDL Number: MFCD01631532 InChI Key: UFPBMVRONDLOGK-UHFFFAOYSA-N Synonym: 3-fluoro-p-tolualdehyde,3-fluoro-4-methyl-benzaldehyde,benzaldehyde, 3-fluoro-4-methyl-9ci,pubchem1454,acmc-1c3kx,ksc494i5j,3-fluoranyl-4-methyl-benzaldehyde,3-fluoro-4-methylbenzaldehyde,benzaldehyde, 3-fluoro-4-methyl PubChem CID: 2774574 IUPAC Name: 3-fluoro-4-methylbenzaldehyde SMILES: CC1=C(C=C(C=C1)C=O)F