missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-methoxybenzaldehyde, 98%

Catalog No. AAB2382706
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2382706 5 g
AAB2382703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2382706 Supplier Thermo Scientific Chemicals Supplier No. B2382706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 351-54-2 | C8H7FO2 | 154.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 351-54-2
Molecular Formula C8H7FO2
Molecular Weight (g/mol) 154.14
MDL Number MFCD00003349
InChI Key SOQCZBSZZLWDGU-UHFFFAOYSA-N
Synonym 3-fluoro-p-anisaldehyde, 3-fluoro-4-methoxy-benzaldehyde, 3-fluoro-4-anisaldehyde, benzaldehyde, 3-fluoro-4-methoxy, 3-fluoro-4-methoxy benzaldehyde, 3-fluoroanisaldehyde, 3-fluoro-anisaldehyde, pubchem1448, acmc-209idn, 2-fluoro-4-formylanisole
PubChem CID 67696
IUPAC Name 3-fluoro-4-methoxybenzaldehyde
SMILES COC1=C(C=C(C=C1)C=O)F

Specifications

Melting Point 34°C to 35°C
Boiling Point 129°C to 132°C (11 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
Beilstein 1942621
Sensitivity Air Sensitive
Formula Weight 154.14
Percent Purity 98%
Chemical Name or Material 3-Fluoro-4-methoxybenzaldehyde
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 206-514-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only