missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Fluoro-4-(4-morpholinylmethyl)benzeneboronic acid pinacol ester, 96%
CAS: 1073354-74-1 | C17H25BFNO3 | 321.20 g/mol
$128.51 - $397.19
Chemical Identifiers
| CAS | 1073354-74-1 |
|---|---|
| Molecular Formula | C17H25BFNO3 |
| Molecular Weight (g/mol) | 321.20 |
| MDL Number | MFCD09027076 |
| InChI Key | QINFDPZSZHPUSC-UHFFFAOYSA-N |
| Synonym | 4-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzyl morpholine, 3-fluoro-4-n-morpholinomethyl phenylboronic acid, pinacol ester, 4-2-fluoro-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl morpholine, 3-fluoro-4-n-morpholinomethyl phenylboronic acid pinacol ester, 4-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl morpholine, 3-fluoro-4-morpholin-4-ylmethyl benzeneboronic acid pinacol ester, 3-fluoro-4-morpholin-4-ylmethyl benzeneboronic acid, pinacol ester, 3-fluoro-4-morpholin-4-ylmethyl benzene boronic acid, pinacol ester, 4-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl benzyl morpholine |
| PubChem CID | 43811063 |
| IUPAC Name | 4-[[2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl]morpholine |
| SMILES | CC1(C)OB(OC1(C)C)C1=CC(F)=C(CN2CCOCC2)C=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH62530MD
|
Thermo Scientific Chemicals
H62530MD |
250 mg |
Each for $128.51
|
|
|||||
|
AAH6253003
|
Thermo Scientific Chemicals
H6253003 |
1 g |
Each for $397.19
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 1073354-74-1 | |
| 321.20 | |
| QINFDPZSZHPUSC-UHFFFAOYSA-N | |
| 43811063 | |
| CC1(C)OB(OC1(C)C)C1=CC(F)=C(CN2CCOCC2)C=C1 |
| C17H25BFNO3 | |
| MFCD09027076 | |
| 4-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzyl morpholine, 3-fluoro-4-n-morpholinomethyl phenylboronic acid, pinacol ester, 4-2-fluoro-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl morpholine, 3-fluoro-4-n-morpholinomethyl phenylboronic acid pinacol ester, 4-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl morpholine, 3-fluoro-4-morpholin-4-ylmethyl benzeneboronic acid pinacol ester, 3-fluoro-4-morpholin-4-ylmethyl benzeneboronic acid, pinacol ester, 3-fluoro-4-morpholin-4-ylmethyl benzene boronic acid, pinacol ester, 4-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl benzyl morpholine | |
| 4-[[2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl]morpholine |
Specifications
| 1073354-74-1 | |
| C17H25BFNO3 | |
| 250 mg | |
| QINFDPZSZHPUSC-UHFFFAOYSA-N | |
| 4-[[2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl]morpholine | |
| 43811063 | |
| 96% | |
| 3-Fluoro-4-(4-morpholinylmethyl)benzeneboronic acid pinacol ester |
| 35°C to 40°C | |
| MFCD09027076 | |
| 4-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzyl morpholine, 3-fluoro-4-n-morpholinomethyl phenylboronic acid, pinacol ester, 4-2-fluoro-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl morpholine, 3-fluoro-4-n-morpholinomethyl phenylboronic acid pinacol ester, 4-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl morpholine, 3-fluoro-4-morpholin-4-ylmethyl benzeneboronic acid pinacol ester, 3-fluoro-4-morpholin-4-ylmethyl benzeneboronic acid, pinacol ester, 3-fluoro-4-morpholin-4-ylmethyl benzene boronic acid, pinacol ester, 4-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl benzyl morpholine | |
| CC1(C)OB(OC1(C)C)C1=CC(F)=C(CN2CCOCC2)C=C1 | |
| 321.20 | |
| 321.19 | |
| Solid |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only