missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-(4-morpholinylmethyl)benzeneboronic acid pinacol ester, 96%

CAS: 1073354-74-1 | C17H25BFNO3 | 321.20 g/mol

Supplier:  Thermo Scientific Chemicals H6253003

 View more versions of this product

Catalog No. AAH6253003

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Fluoro-4-(4-morpholinylmethyl)benzeneboronic acid pinacol ester
35°C to 40°C
MFCD09027076
4-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzyl morpholine, 3-fluoro-4-n-morpholinomethyl phenylboronic acid, pinacol ester, 4-2-fluoro-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl morpholine, 3-fluoro-4-n-morpholinomethyl phenylboronic acid pinacol ester, 4-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl morpholine, 3-fluoro-4-morpholin-4-ylmethyl benzeneboronic acid pinacol ester, 3-fluoro-4-morpholin-4-ylmethyl benzeneboronic acid, pinacol ester, 3-fluoro-4-morpholin-4-ylmethyl benzene boronic acid, pinacol ester, 4-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl benzyl morpholine
CC1(C)OB(OC1(C)C)C1=CC(F)=C(CN2CCOCC2)C=C1
321.20
321.19
Solid
1073354-74-1
C17H25BFNO3
1 g
QINFDPZSZHPUSC-UHFFFAOYSA-N
4-[[2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl]morpholine
43811063
96%

Chemical Identifiers

1073354-74-1
321.20
QINFDPZSZHPUSC-UHFFFAOYSA-N
43811063
CC1(C)OB(OC1(C)C)C1=CC(F)=C(CN2CCOCC2)C=C1
C17H25BFNO3
MFCD09027076
4-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzyl morpholine, 3-fluoro-4-n-morpholinomethyl phenylboronic acid, pinacol ester, 4-2-fluoro-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl morpholine, 3-fluoro-4-n-morpholinomethyl phenylboronic acid pinacol ester, 4-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl morpholine, 3-fluoro-4-morpholin-4-ylmethyl benzeneboronic acid pinacol ester, 3-fluoro-4-morpholin-4-ylmethyl benzeneboronic acid, pinacol ester, 3-fluoro-4-morpholin-4-ylmethyl benzene boronic acid, pinacol ester, 4-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl benzyl morpholine
4-[[2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl]morpholine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only