missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-2-nitrophenol 98.0+%, TCI America™

Catalog No. F09475G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F09475G 5 g
F09471G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F09475G Supplier TCI America Supplier No. F09475G

Chemical Identifiers

CAS 385-01-3
Molecular Formula C6H4FNO3
Molecular Weight (g/mol) 157.1
MDL Number MFCD07368753
InChI Key GAWNBKUTBVLIPL-UHFFFAOYSA-N
Synonym 3-fluoro-2-nitro-phenol, 2-nitro-3-fluorophenol, phenol, 3-fluoro-2-nitro, 3-fluoronitro-phenol, pubchem4124, acmc-209j0i, ksc497m1h
PubChem CID 2782777
IUPAC Name 3-fluoro-2-nitrophenol
SMILES C1=CC(=C(C(=C1)F)[N+](=O)[O-])O

Specifications

Melting Point 38°C
Quantity 5 g
Formula Weight 157.10
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Fluoro-2-nitrophenol
TSCA No
Recommended Storage Refrigerator