accessibility menu, dialog, popup

UserName

FNOOHO

CAS RN 385-01-3

IUPAC Name: 3-fluoro-2-nitrophenol
Synonyms: 3-fluoro-2-nitrophenol
Molecular Weight (g/mol): 157.1
Molecular Formula: C6H4FNO3
InChi Key: GAWNBKUTBVLIPL-UHFFFAOYSA-N
SMILES: OC1=C(C(F)=CC=C1)[N+]([O-])=O
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)