missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-2-methylbenzeneboronic acid, 98%, Thermo Scientific Chemicals

Catalog No. AAH5275203
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5275203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5275203 Supplier Thermo Scientific Chemicals Supplier No. H5275203
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 163517-61-1 | Molecular Formula: C7H8BFO2 | Molecular Weight: 153.947 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 163517-61-1
Molecular Formula C7H8BFO2
Molecular Weight (g/mol) 153.947
MDL Number MFCD07368679
InChI Key NYBIUWJUWTUGFV-UHFFFAOYSA-N
Synonym 3-fluoro-2-methylphenyl boronic acid, 2-methyl-3-fluoro-phenylboronic acid, 3-fluoro-2-methylbenzeneboronic acid, 3-fluoro-o-tolylboronic acid, boronic acid, 3-fluoro-2-methylphenyl, boronic acid,b-3-fluoro-2-methylphenyl, 3-fluoro-2-methyl-phenyl boronic acid, 2-borono-6-fluorotoluene, acmc-1c5qk, 2-methyl-3-fluorophenylboronic acid
PubChem CID 2783191
IUPAC Name (3-fluoro-2-methylphenyl)boronic acid
SMILES B(C1=C(C(=CC=C1)F)C)(O)O

Specifications

Melting Point 142°C to 144°C
Quantity 1 g
Formula Weight 153.95
Percent Purity 98%
Chemical Name or Material 3-Fluoro-2-methylbenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only