missing translation for 'onlineSavingsMsg'
Learn More

3-Ethyltoluene 98.0+%, TCI America™

Supplier:  TCI America E018525ML

 View more versions of this product

Catalog No. E018525ML


Only null left

Description

Description

Specifications

Specifications

3-Ethyltoluene
Liquid
MFCD00009259
Yellow
3-Methylethylbenzene
CCC1=CC=CC(C)=C1
120.20
12100
120.20
620-14-4
C9H12
3295
160°C
ZLCSFXXPPANWQY-UHFFFAOYSA-N
1-ethyl-3-methylbenzene
25 mL
CHEBI:77512
≥98.0% (GC)

Chemical Identifiers

620-14-4
120.20
ZLCSFXXPPANWQY-UHFFFAOYSA-N
12100
1-ethyl-3-methylbenzene
C9H12
MFCD00009259
3-Methylethylbenzene
CHEBI:77512
CCC1=CC=CC(C)=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0015

RTECSNumber : DA0712000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions