missing translation for 'onlineSavingsMsg'
Learn More

3-Ethyltoluene 98.0+%, TCI America™

Catalog No. p-7128884
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E01855ML 5 mL
E018525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E01855ML Supplier TCI America Supplier No. E01855ML
Only null left

Chemical Identifiers

CAS 620-14-4
Molecular Formula C9H12
Molecular Weight (g/mol) 120.20
MDL Number MFCD00009259
InChI Key ZLCSFXXPPANWQY-UHFFFAOYSA-N
Synonym 3-Methylethylbenzene
PubChem CID 12100
ChEBI CHEBI:77512
IUPAC Name 1-ethyl-3-methylbenzene
SMILES CCC1=CC=CC(C)=C1

Specifications

Physical Form Liquid
UN Number 3295
Color Yellow
Boiling Point 160°C
Quantity 5 mL
Formula Weight 120.20
Percent Purity ≥98.0% (GC)
Chemical Name or Material 3-Ethyltoluene
EINECSNumber (3)-0015
RTECSNumber DA0712000
TSCA No