missing translation for 'onlineSavingsMsg'
Learn More

3-Ethylamino-p-cresol 92.0+%, TCI America™

Catalog No. E025225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E025225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E025225G Supplier TCI America Supplier No. E025225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 120-37-6
Molecular Formula C9H13NO
Molecular Weight (g/mol) 151.21
MDL Number MFCD00002382
InChI Key CTGSQPRDMHCIMM-UHFFFAOYSA-N
Synonym 3-ethylamino-4-methylphenol, 3-ethylamino-p-cresol, phenol, 3-ethylamino-4-methyl, ccris 4639, 3-ethylamino-4-methyl-phenol, unii-8002qme89h, 3-ethylamino-4-cresol, 3-ethylamino p-cresol, dsstox_cid_5269
PubChem CID 8428
IUPAC Name 3-(ethylamino)-4-methylphenol
SMILES CCNC1=CC(O)=CC=C1C

Specifications

Quantity 25 g
Formula Weight 151.21
Percent Purity ≥92.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Ethylamino-p-cresol
RTECSNumber SL4105000
TSCA Yes