accessibility menu, dialog, popup

UserName

CAS RN 120-37-6

IUPAC Name: 3-(ethylamino)-4-methylphenol
Synonyms: 3-(ethylamino)-4-methylphenol
Molecular Weight (g/mol): 151.21
Molecular Formula: C9H13NO
InChi Key: CTGSQPRDMHCIMM-UHFFFAOYSA-N
SMILES: CCNC1=CC(O)=CC=C1C
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Boiling Point 
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)