missing translation for 'onlineSavingsMsg'
Learn More

3-Ethylamino-4-methylphenol, tech. 90%

Catalog No. p-7046010
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1276806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1276806 Supplier Thermo Scientific Chemicals Supplier No. L1276806

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 120-37-6 | C9H13NO | 151.21 g/mol

3-Ethylamino-4-methylphenol was used in the synthesis of rhodol by undergoing condensation reaction with 2-(2,4-dihydroxybenzoyl)benzoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Ethylamino-4-methylphenol was used in the synthesis of rhodol by undergoing condensation reaction with 2-(2,4-dihydroxybenzoyl)benzoic acid.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 120-37-6
Molecular Formula C9H13NO
Molecular Weight (g/mol) 151.21
MDL Number MFCD00002382
InChI Key CTGSQPRDMHCIMM-UHFFFAOYSA-N
Synonym 3-ethylamino-4-methylphenol, 3-ethylamino-p-cresol, phenol, 3-ethylamino-4-methyl, ccris 4639, 3-ethylamino-4-methyl-phenol, unii-8002qme89h, 3-ethylamino-4-cresol, 3-ethylamino p-cresol, dsstox_cid_5269
PubChem CID 8428
IUPAC Name 3-(ethylamino)-4-methylphenol
SMILES CCNC1=CC(O)=CC=C1C

Specifications

Melting Point 84°C to 92°C
Boiling Point 135°C to 140°C (3 mmHg)
Quantity 5 g
Beilstein 3241548
Solubility Information Insoluble in water.
Formula Weight 151.21
Percent Purity 90%
Grade Technical
Chemical Name or Material 3-Ethylamino-4-methylphenol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 204-391-0
RTECSNumber SL4105000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only