missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Ethyl (-)-Thiazolidine-3,4-dicarboxylate 98.0+%, TCI America™
Supplier: TCI America T197425G
Specifications
| 3-Ethyl (-)-Thiazolidine-3,4-dicarboxylate | |
| 98°C | |
| C7H11NO4S | |
| 25 g | |
| XBJWOGLKABXFJE-UHFFFAOYSA-N | |
| 3-ethoxycarbonyl-1,3-thiazolidine-4-carboxylic acid | |
| 11615393 | |
| ≥98.0% (T) |
| 127657-29-8 | |
| White-Yellow | |
| MFCD04117921 | |
| (-)-3-Carbethoxythiazolidine-4-carboxylic Acid, (-)-3-Ethoxycarbonylthiazolidine-4-carboxylic Acid, (-)-Thiazolidine-3,4-dicarboxylic Acid 3-Ethyl Ester | |
| CCOC(=O)N1CSCC1C(=O)O | |
| 205.228 | |
| 205.23 | |
| Crystalline Powder |
Chemical Identifiers
| 127657-29-8 | |
| 205.228 | |
| XBJWOGLKABXFJE-UHFFFAOYSA-N | |
| 11615393 | |
| CCOC(=O)N1CSCC1C(=O)O |
| C7H11NO4S | |
| MFCD04117921 | |
| (-)-3-Carbethoxythiazolidine-4-carboxylic Acid, (-)-3-Ethoxycarbonylthiazolidine-4-carboxylic Acid, (-)-Thiazolidine-3,4-dicarboxylic Acid 3-Ethyl Ester | |
| 3-ethoxycarbonyl-1,3-thiazolidine-4-carboxylic acid |
Safety and Handling
TSCA : No