accessibility menu, dialog, popup

UserName

CAS RN 127657-29-8

IUPAC Name: 3-(ethoxycarbonyl)-1,3-thiazolidine-4-carboxylic acid
Synonyms: 3-(ethoxycarbonyl)-1,3-thiazolidine-4-carboxylic acid
Molecular Weight (g/mol): 205.23
Molecular Formula: C7H11NO4S
InChi Key: XBJWOGLKABXFJE-UHFFFAOYNA-N
SMILES: CCOC(=O)N1CSCC1C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results