missing translation for 'onlineSavingsMsg'
Learn More

3-Ethyl (-)-Thiazolidine-3,4-dicarboxylate 98.0+%, TCI America™

Catalog No. T19745G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T19745G 5 g
T197425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T19745G Supplier TCI America Supplier No. T19745G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 127657-29-8
Molecular Formula C7H11NO4S
Molecular Weight (g/mol) 205.228
MDL Number MFCD04117921
InChI Key XBJWOGLKABXFJE-UHFFFAOYSA-N
Synonym (-)-3-Carbethoxythiazolidine-4-carboxylic Acid, (-)-3-Ethoxycarbonylthiazolidine-4-carboxylic Acid, (-)-Thiazolidine-3,4-dicarboxylic Acid 3-Ethyl Ester
PubChem CID 11615393
IUPAC Name 3-ethoxycarbonyl-1,3-thiazolidine-4-carboxylic acid
SMILES CCOC(=O)N1CSCC1C(=O)O

Specifications

Melting Point 98°C
Color White-Yellow
Quantity 5 g
Formula Weight 205.23
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Ethyl (-)-Thiazolidine-3,4-dicarboxylate