missing translation for 'onlineSavingsMsg'
Learn More

3-Ethoxyphenol, 98%

Catalog No. AAB2426622
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2426622 100 g
AAB2426606 5 g
AAB2426614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2426622 Supplier Thermo Scientific Chemicals Supplier No. B2426622
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 621-34-1 | C8H10O2 | 138.166 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 621-34-1
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.166
MDL Number MFCD00016450
InChI Key VBIKLMJHBGFTPV-UHFFFAOYSA-N
Synonym m-ethoxyphenol, phenol, 3-ethoxy, phenol, m-ethoxy, resorcinol monoethyl ether, 3-ethoxy-phenol, unii-iky4o3474a, 3-ethyloxy phenol, pubchem9510, acmc-209mzu, phenol, m-ethoxy-8ci
PubChem CID 69306
IUPAC Name 3-ethoxyphenol
SMILES CCOC1=CC=CC(=C1)O

Specifications

Melting Point 28°C to 31°C
Density 1.07
Boiling Point 247°C
Flash Point >93°C (199°F)
Odor Phenol-like
Refractive Index 1.541
Quantity 100 g
Formula Weight 138.17
Percent Purity 98%
Chemical Name or Material 3-Ethoxyphenol
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 210-681-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only