accessibility menu, dialog, popup

UserName

CAS RN 621-34-1

IUPAC Name: 3-ethoxyphenol
Synonyms: M-ethoxyphenol
Molecular Weight (g/mol): 138.17
Molecular Formula: C8H10O2
InChi Key: VBIKLMJHBGFTPV-UHFFFAOYSA-N
SMILES: CCOC1=CC=CC(O)=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (5)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)