missing translation for 'onlineSavingsMsg'
Learn More

3-Ethoxybenzonitrile, 95%

Catalog No. AAA1390814
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1390814 25 g
AAA1390806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1390814 Supplier Thermo Scientific Chemicals Supplier No. A1390814
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 25117-75-3 | C9H9NO | 147.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 25117-75-3
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.18
MDL Number MFCD00001805
InChI Key PFNAUBYHTWDKBO-UHFFFAOYSA-N
Synonym benzonitrile, 3-ethoxy, benzonitrile, m-ethoxy, m-ethoxy benzonitrile, acmc-20an4n, 3-ethoxybenzenecarbonitrile, 3-ethoxybenzoic acid nitrile
PubChem CID 91283
IUPAC Name 3-ethoxybenzonitrile
SMILES CCOC1=CC=CC(=C1)C#N

Specifications

Density 1.047
Boiling Point 241°C to 244°C
Flash Point >110°C (230°F)
Refractive Index 1.529
Quantity 25 g
UN Number UN3276
Formula Weight 147.18
Percent Purity 95%
Chemical Name or Material 3-Ethoxybenzonitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
EINECSNumber 246-632-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only