accessibility menu, dialog, popup

UserName

CH3ON

CAS RN 25117-75-3

IUPAC Name: 3-ethoxybenzonitrile
Synonyms: 3-ethoxybenzonitrile
Molecular Weight (g/mol): 147.18
Molecular Formula: C9H9NO
InChi Key: PFNAUBYHTWDKBO-UHFFFAOYSA-N
SMILES: CCOC1=CC=CC(=C1)C#N
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results