Learn More
3-Ethoxybenzoic acid, 98+%, Thermo Scientific Chemicals
$192.42 - $192.42
Chemical Identifiers
| CAS | 621-51-2 |
|---|---|
| Molecular Formula | C9H10O3 |
| Molecular Weight (g/mol) | 166.176 |
| MDL Number | MFCD00016517 |
| InChI Key | DTFQMPQJMDEWKJ-UHFFFAOYSA-N |
| Synonym | 3-ethoxybenzoate, m-ethoxybenzoic acid, benzoic acid, 3-ethoxy, 3-ethoxy-benzoic acid, benzoic acid, m-ethoxy, 3-ethoxybenzoicacid, 3-ethyloxy benzoic acid, 3-ethoxybenzoic acid |
| PubChem CID | 12126 |
| ChEBI | CHEBI:27990 |
| IUPAC Name | 3-ethoxybenzoic acid |
| SMILES | CCOC1=CC=CC(=C1)C(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL0572109
|
Thermo Scientific Chemicals
L0572109 |
10 g |
Each for $192.42
|
|
|||||
Description
3-Ethoxybenzoic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Ethoxybenzoic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in well sealed container.
Chemical Identifiers
| 621-51-2 | |
| 166.176 | |
| DTFQMPQJMDEWKJ-UHFFFAOYSA-N | |
| 12126 | |
| 3-ethoxybenzoic acid |
| C9H10O3 | |
| MFCD00016517 | |
| 3-ethoxybenzoate, m-ethoxybenzoic acid, benzoic acid, 3-ethoxy, 3-ethoxy-benzoic acid, benzoic acid, m-ethoxy, 3-ethoxybenzoicacid, 3-ethyloxy benzoic acid, 3-ethoxybenzoic acid | |
| CHEBI:27990 | |
| CCOC1=CC=CC(=C1)C(=O)O |
Specifications
| 621-51-2 | |
| C9H10O3 | |
| 10 g | |
| 3-ethoxybenzoate, m-ethoxybenzoic acid, benzoic acid, 3-ethoxy, 3-ethoxy-benzoic acid, benzoic acid, m-ethoxy, 3-ethoxybenzoicacid, 3-ethyloxy benzoic acid, 3-ethoxybenzoic acid | |
| CCOC1=CC=CC(=C1)C(=O)O | |
| 166.176 | |
| CHEBI:27990 | |
| ≥98% |
| 135°C to 137°C | |
| MFCD00016517 | |
| 509159 | |
| DTFQMPQJMDEWKJ-UHFFFAOYSA-N | |
| 3-ethoxybenzoic acid | |
| 12126 | |
| 166.18 | |
| 3-Ethoxybenzoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 210-690-7
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only