accessibility menu, dialog, popup

UserName

CAS RN 621-51-2

IUPAC Name: 3-ethoxybenzoic acid
Synonyms: 3-ethoxybenzoic acid
Molecular Weight (g/mol): 166.18
Molecular Formula: C9H10O3
InChi Key: DTFQMPQJMDEWKJ-UHFFFAOYSA-N
SMILES: CCOC1=CC=CC(=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results