missing translation for 'onlineSavingsMsg'
Learn More

3-Ethoxybenzoic acid, 98+%, Thermo Scientific Chemicals

Catalog No. AAL0572109
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0572109 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0572109 Supplier Thermo Scientific Chemicals Supplier No. L0572109
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 621-51-2 | Molecular Formula: C9H10O3 | Molecular Weight: 166.176 g/mol

3-Ethoxybenzoic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Ethoxybenzoic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in well sealed container.

Chemical Identifiers

CAS 621-51-2
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00016517
InChI Key DTFQMPQJMDEWKJ-UHFFFAOYSA-N
Synonym 3-ethoxybenzoate, m-ethoxybenzoic acid, benzoic acid, 3-ethoxy, 3-ethoxy-benzoic acid, benzoic acid, m-ethoxy, 3-ethoxybenzoicacid, 3-ethyloxy benzoic acid, 3-ethoxybenzoic acid
PubChem CID 12126
ChEBI CHEBI:27990
IUPAC Name 3-ethoxybenzoic acid
SMILES CCOC1=CC=CC(=C1)C(=O)O

Specifications

Melting Point 135°C to 137°C
Quantity 10 g
Beilstein 509159
Formula Weight 166.18
Percent Purity ≥98%
Chemical Name or Material 3-Ethoxybenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-690-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only