Learn More
3-Ethoxy-5-(trifluoromethyl)benzeneboronic acid, 98%, Thermo Scientific Chemicals
$349.53 - $349.53
Chemical Identifiers
| CAS | 871332-96-6 |
|---|---|
| Molecular Formula | C9H10BF3O3 |
| Molecular Weight (g/mol) | 233.98 |
| MDL Number | MFCD08056361 |
| InChI Key | ATZRXKHRSOTBAM-UHFFFAOYSA-N |
| Synonym | 3-ethoxy-5-trifluoromethyl phenylboronic acid, 3-ethoxy-5-trifluoromethyl benzeneboronic acid, boronic acid,b-3-ethoxy-5-trifluoromethyl phenyl, 3-ethoxy-5-trifluoromethylphenylboronic acid, 3-ethoxy-5-trifluoromethyl phenyl boronic acid, acmc-209qhp, 3-ethoxy-5-trifluoromethyl benzeneboronicacid |
| PubChem CID | 44717589 |
| IUPAC Name | [3-ethoxy-5-(trifluoromethyl)phenyl]boronic acid |
| SMILES | CCOC1=CC(=CC(=C1)C(F)(F)F)B(O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Qty | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Qty | |||||
|
AAH5298903
|
Thermo Scientific Chemicals
H5298903 |
1 g |
N/A
|
|
|||||
Description
Reactant involved in suzuki-miyaura cross-coupling reactions, aerobic oxidative cross-coupling, rhodium-catalyzed addition reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsReactant involved in suzuki-miyaura cross-coupling reactions, aerobic oxidative cross-coupling, rhodium-catalyzed addition reactions.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Store at 4°C.
Chemical Identifiers
| 871332-96-6 | |
| 233.98 | |
| ATZRXKHRSOTBAM-UHFFFAOYSA-N | |
| 44717589 | |
| CCOC1=CC(=CC(=C1)C(F)(F)F)B(O)O |
| C9H10BF3O3 | |
| MFCD08056361 | |
| 3-ethoxy-5-trifluoromethyl phenylboronic acid, 3-ethoxy-5-trifluoromethyl benzeneboronic acid, boronic acid,b-3-ethoxy-5-trifluoromethyl phenyl, 3-ethoxy-5-trifluoromethylphenylboronic acid, 3-ethoxy-5-trifluoromethyl phenyl boronic acid, acmc-209qhp, 3-ethoxy-5-trifluoromethyl benzeneboronicacid | |
| [3-ethoxy-5-(trifluoromethyl)phenyl]boronic acid |
Specifications
| 871332-96-6 | |
| C9H10BF3O3 | |
| 1 g | |
| Slightly soluble in water. | |
| CCOC1=CC(=CC(=C1)C(F)(F)F)B(O)O | |
| 233.98 | |
| 233.98 | |
| 3-Ethoxy-5-(trifluoromethyl)benzeneboronic acid |
| 173°C to 176°C | |
| MFCD08056361 | |
| 3-ethoxy-5-trifluoromethyl phenylboronic acid, 3-ethoxy-5-trifluoromethyl benzeneboronic acid, boronic acid,b-3-ethoxy-5-trifluoromethyl phenyl, 3-ethoxy-5-trifluoromethylphenylboronic acid, 3-ethoxy-5-trifluoromethyl phenyl boronic acid, acmc-209qhp, 3-ethoxy-5-trifluoromethyl benzeneboronicacid | |
| ATZRXKHRSOTBAM-UHFFFAOYSA-N | |
| [3-ethoxy-5-(trifluoromethyl)phenyl]boronic acid | |
| 44717589 | |
| 98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only