missing translation for 'onlineSavingsMsg'
Learn More

3-Ethoxy-4-methoxybenzaldehyde, 99%

Catalog No. p-7027759
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2476206 5 g
AAB2476214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2476206 Supplier Thermo Scientific Chemicals Supplier No. B2476206

CAS: 1131-52-8 | C10H12O3 | 180.203 g/mol

3-Ethoxy-4-methoxybenzaldehyde is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Ethoxy-4-methoxybenzaldehyde is used as a pharmaceutical intermediate.

Solubility
Soluble in methanol.

Notes
Air sensitive. Store in a tightly sealed container. Store away from oxidizing agents, air.

Chemical Identifiers

CAS 1131-52-8
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD00010128
InChI Key VAMZHXWLGRQSJS-UHFFFAOYSA-N
Synonym 3-ethoxy-4-methoxy-benzaldehyde, benzaldehyde, 3-ethoxy-4-methoxy, pubchem13135, 3-ethoxy-p-anisaldehyde, acmc-1ca82, 3-ethoxy-4methoxybenzaldehyde, 3ethoxy-4-methoxybenzaldehyde, ksc183c6j, 3-ethoxy-4-methoxybenzaldehyd, 4-methoxy-3-ethoxybenzaldehyde
PubChem CID 244728
IUPAC Name 3-ethoxy-4-methoxybenzaldehyde
SMILES CCOC1=C(C=CC(=C1)C=O)OC

Specifications

Melting Point 51°C to 53°C
Quantity 5 g
Sensitivity Air Sensitive
Solubility Information Soluble in methanol.
Formula Weight 180.2
Percent Purity 99%
Chemical Name or Material 3-Ethoxy-4-methoxybenzaldehyde
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only