Learn More
3-(Dimethylsulfamoyl)benzeneboronic acid, 98%
CAS: 871329-59-8 | C8H12BNO4S | 229.057 g/mol
$197.71 - $566.66
Chemical Identifiers
| CAS | 871329-59-8 |
|---|---|
| Molecular Formula | C8H12BNO4S |
| Molecular Weight (g/mol) | 229.057 |
| MDL Number | MFCD07363747 |
| InChI Key | SGERXKCDWJPIOS-UHFFFAOYSA-N |
| Synonym | 3-n,n-dimethylsulfamoyl phenyl boronic acid, n,n-dimethyl 3-boronobenzenesulfonamide, 3-dimethylsulfamoyl phenyl boronic acid, 3-n,n-dimethylsulphonamido benzeneboronic acid, 3-dimethylsulfamoyl phenylboronic acid, 3-dimethylsulfamoyl benzeneboronic acid, 3-n,n-dimethylsulfonamidophenyl boronic acid, boronic acid, 3-dimethylamino sulfonyl phenyl, boronic acid,b-3-dimethylamino sulfonyl phenyl, acmc-209qg4 |
| PubChem CID | 44119634 |
| IUPAC Name | [3-(dimethylsulfamoyl)phenyl]boronic acid |
| SMILES | B(C1=CC(=CC=C1)S(=O)(=O)N(C)C)(O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH52838MD
|
Thermo Scientific Chemicals
H52838MD |
250 mg |
N/A
|
|
|||||
|
AAH5283803
|
Thermo Scientific Chemicals
H5283803 |
1 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 871329-59-8 | |
| 229.057 | |
| SGERXKCDWJPIOS-UHFFFAOYSA-N | |
| 44119634 | |
| B(C1=CC(=CC=C1)S(=O)(=O)N(C)C)(O)O |
| C8H12BNO4S | |
| MFCD07363747 | |
| 3-n,n-dimethylsulfamoyl phenyl boronic acid, n,n-dimethyl 3-boronobenzenesulfonamide, 3-dimethylsulfamoyl phenyl boronic acid, 3-n,n-dimethylsulphonamido benzeneboronic acid, 3-dimethylsulfamoyl phenylboronic acid, 3-dimethylsulfamoyl benzeneboronic acid, 3-n,n-dimethylsulfonamidophenyl boronic acid, boronic acid, 3-dimethylamino sulfonyl phenyl, boronic acid,b-3-dimethylamino sulfonyl phenyl, acmc-209qg4 | |
| [3-(dimethylsulfamoyl)phenyl]boronic acid |
Specifications
| 871329-59-8 | |
| C8H12BNO4S | |
| 250 mg | |
| SGERXKCDWJPIOS-UHFFFAOYSA-N | |
| [3-(dimethylsulfamoyl)phenyl]boronic acid | |
| 44119634 | |
| 98% |
| 88°C to 92°C | |
| MFCD07363747 | |
| 3-n,n-dimethylsulfamoyl phenyl boronic acid, n,n-dimethyl 3-boronobenzenesulfonamide, 3-dimethylsulfamoyl phenyl boronic acid, 3-n,n-dimethylsulphonamido benzeneboronic acid, 3-dimethylsulfamoyl phenylboronic acid, 3-dimethylsulfamoyl benzeneboronic acid, 3-n,n-dimethylsulfonamidophenyl boronic acid, boronic acid, 3-dimethylamino sulfonyl phenyl, boronic acid,b-3-dimethylamino sulfonyl phenyl, acmc-209qg4 | |
| B(C1=CC(=CC=C1)S(=O)(=O)N(C)C)(O)O | |
| 229.057 | |
| 229.06 | |
| 3-(Dimethylsulfamoyl)benzeneboronic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only